Learn More
Thermo Scientific™ 6-Place Fixed-Angle Highconic™ Rotor Adapters
Use these rotor adapters to connect with round or conical-bottom centrifuge tubes and microtubes tubes up to 50mL.
Brand: Thermo Scientific™ 76002905
Requires : 50mL conical adapter 75-007-556 requires rubber pads (supplied with buckets). Remove pads when using all other size adapters.
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Descrizione
Use these 6-Place Fixed-Angle Highconic™ Rotor Adapters to connect with centrifuge tubes up to 50mL. The adapters accommodate round or conical-bottom tubes and microtubes and work with HiGhConic™ fixed angle rotors.
The adapters accommodate round- or conical-bottom tubes and microtubes.
Order Info
Adapters are sold individually; order correct quantity to equip rotor.
Specifica
Microtube | |
HiGhConic™ fixed angle rotor | |
1.5 mL |
Offerte speciali
-
Multiple Manufacturers
The Academic Best Buy Book 2025
Browse exclusive prices for our academic customersPromo Code: 25081Expires: 2025-12-31
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.