missing translation for 'onlineSavingsMsg'
Learn More
7-Bromo-2,3-dihydrothieno[3,4-b][1,4]dioxine-5-carbaldehyde, 97%, Thermo Scientific™ Product Code.: 10602865

7-Bromo-2,3-dihydrothieno[3,4-b][1,4]dioxine-5-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10602865
1 g
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy

Product Code. 10602865

Brand: Thermo Scientific Maybridge CC58104DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 852054-42-3
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C7H5BrO3S
MDL Number MFCD08690306
Quantity 1 g
Synonym 7-bromo-2,3-dihydrothieno 3,4-b 1,4 dioxine-5-carbaldehyde, 7-bromo-2h,3h-thieno 3,4-b 1,4 dioxine-5-carbaldehyde, 7-bromo-2,3-dihydrothieno 3,4-b 1,4 dioxine-5-carboxaldehyde, 2,3-dihydro-7-bromothieno 3,4-b 1,4 dioxin-5-carbaldehyde, 7-bromo-2,3-dihydrothieno 3,4-b-1,4-dioxin-5-carbaldehyde, 7-bromo-2h,3h-thiopheno 3,4-e 1,4-dioxane-5-carbaldehyde, thieno 3,4-b-1,4-dioxin-5-carboxaldehyde,7-bromo-2,3-dihydro
InChI Key YEKBJVHBVICUOZ-UHFFFAOYSA-N
SMILES C1COC2=C(SC(=C2O1)C=O)Br
IUPAC Name 5-bromo-2,3-dihydrothieno[3,4-b][1,4]dioxine-7-carbaldehyde
Molecular Weight (g/mol) 249.078
PubChem CID 11391008
Formula Weight 249.08
Percent Purity 97%
Chemical Name or Material 7-Bromo-2,3-dihydrothieno[3,4-b][1,4]dioxine-5-carbaldehyde
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
7-Bromo-2,3-dihydrothieno[3,4-b][1,4]dioxine-5-carbaldehyde, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.