Learn More
JRI Maxant™ 7-Day Thermographer
1940.09€
Specifications
Description Temp. Range: -35°C to 15°C
View More Specs
Products 2
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
12668241
Encompass
View accessory products View Documents
JRI Maxant™
00006
Temp. Range: -35°C to 15°C
1940.09€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 09-01-2025
to see stock.
Add to basket
12688241
Encompass
View accessory products View Documents
JRI Maxant™
00060
Temp. Range: -35°C to 15°C
1940.09€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 31-01-2025
Specifications
Temp. Range: -35°C to 15°C