missing translation for 'onlineSavingsMsg'
Learn More

7-Methoxybenzo[b]furan-2-carboxylic acid, 97%, Thermo Scientific™

Product Code. 11424687
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4790-79-8
Molecular Formula C10H8O4
Molecular Weight (g/mol) 192.17
MDL Number MFCD00060516
InChI Key UOCNTRAAJNWDND-UHFFFAOYSA-N
Synonym 7-methoxybenzofuran-2-carboxylic acid, 7-methoxybenzo b furan-2-carboxylic acid, 2-benzofurancarboxylic acid, 7-methoxy, 7-methoxy-2-benzofurancarboxylic acid, 7-methoxy-benzofuran-2-carboxylic acid, 7-methoxybenzofuran-2-carboxylicacid, acmc-1am7f, ksc236k1t, 7methoxybenzofuran-2-carboxylic acid
PubChem CID 606037
IUPAC Name 7-methoxy-1-benzofuran-2-carboxylic acid
SMILES COC1=CC=CC2=C1OC(=C2)C(=O)O

Specifications

Melting Point 217°C to 220°C
Quantity 1 g
Formula Weight 192.17
Percent Purity 97%
Chemical Name or Material 7-Methoxybenzo[b]furan-2-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'