missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Nitro-1-tetralone, 97%, Thermo Scientific Chemicals
74.30€ - 251.00€
Chemical Identifiers
CAS | 40353-34-2 |
---|---|
Molecular Formula | C10H9NO3 |
Molecular Weight (g/mol) | 191.186 |
MDL Number | MFCD00019661 |
InChI Key | GWAQYWSNCVEJMW-UHFFFAOYSA-N |
Synonym | 7-nitro-1-tetralone, 7-nitrotetralone, 7-nitro-3,4-dihydronaphthalen-1 2h-one, 1 2h-naphthalenone, 3,4-dihydro-7-nitro, 3,4-dihydro-7-nitro-1 2h-naphthalenone, 3,4-dihydro-7-nitronaphthalen-1 2h-one, wln: l66 bvt&j inw, 7-nitro-2,3,4-trihydronaphthalen-1-one, 1 2h-naphthalenone,4-dihydro-7-nitro, 7-nitro-tetralone |
PubChem CID | 38445 |
IUPAC Name | 7-nitro-3,4-dihydro-2H-naphthalen-1-one |
SMILES | C1CC2=C(C=C(C=C2)[N+](=O)[O-])C(=O)C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11301427
|
Thermo Scientific Alfa Aesar
L01853.04 |
2 g |
74.30€
2g |
Estimated Shipment: 30-10-2024
Log in to see stock. |
|||||
11311427
|
Thermo Scientific Alfa Aesar
L01853.09 |
10 g |
251.00€
10g |
Estimated Shipment: 30-10-2024
Log in to see stock. |
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
40353-34-2 | |
191.186 | |
GWAQYWSNCVEJMW-UHFFFAOYSA-N | |
38445 | |
C1CC2=C(C=C(C=C2)[N+](=O)[O-])C(=O)C1 |
C10H9NO3 | |
MFCD00019661 | |
7-nitro-1-tetralone, 7-nitrotetralone, 7-nitro-3,4-dihydronaphthalen-1 2h-one, 1 2h-naphthalenone, 3,4-dihydro-7-nitro, 3,4-dihydro-7-nitro-1 2h-naphthalenone, 3,4-dihydro-7-nitronaphthalen-1 2h-one, wln: l66 bvt&j inw, 7-nitro-2,3,4-trihydronaphthalen-1-one, 1 2h-naphthalenone,4-dihydro-7-nitro, 7-nitro-tetralone | |
7-nitro-3,4-dihydro-2H-naphthalen-1-one |
Specifications
40353-34-2 | |
C10H9NO3 | |
1570515 | |
GWAQYWSNCVEJMW-UHFFFAOYSA-N | |
7-nitro-3,4-dihydro-2H-naphthalen-1-one | |
38445 | |
97% |
103°C to 108°C | |
MFCD00019661 | |
7-nitro-1-tetralone, 7-nitrotetralone, 7-nitro-3,4-dihydronaphthalen-1 2h-one, 1 2h-naphthalenone, 3,4-dihydro-7-nitro, 3,4-dihydro-7-nitro-1 2h-naphthalenone, 3,4-dihydro-7-nitronaphthalen-1 2h-one, wln: l66 bvt&j inw, 7-nitro-2,3,4-trihydronaphthalen-1-one, 1 2h-naphthalenone,4-dihydro-7-nitro, 7-nitro-tetralone | |
C1CC2=C(C=C(C=C2)[N+](=O)[O-])C(=O)C1 | |
191.186 | |
191.19 | |
7-Nitro-1-tetralone |
Safety and Handling
missing translation for 'einecsNumber' : 254-887-6
missing translation for 'rtecsNumber' : QK5025000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only