missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ 700 Series Cemented Needle Custom Syringes

Syringes for customized needles. This series of syringes is ideal for dispensing volumes from 0.5 up to 500μL.

Brand:  Hamilton™ 80608

 View more versions of this product

Product Code. 10743903

  • 170.00€ / Each
Estimated Shipment: 18-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • The original hand-fitted Hamilton™ syringe
  • Stainless steel plunger is manufactured to fit the glass barrel with a tolerance less than three thousandths of a millimeter
  • Unsurpassed syringe life

Order Info

This is a Custom Order. Please consult with your Fisher Sales Representative before ordering.

Specifications

Specifications

Not Autoclavable
Custom
7.75 mm
Microliter Syringe
50°C
10°C
100 μL
Custom
Borosilicate Glass, Stainless Steel
1.46 mm
Ideal for dispensing volumes from 0.5 μL up to 500 μL
2000 psig (137.9 bar)
710 SN
1/Ea.
Custom Length
Cemented Needle Special
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers