missing translation for 'onlineSavingsMsg'
Learn More

8-Bromo-cAMP, sodium salt, Tocris Bioscience™

Product Code. 18055122
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Cell-permeable cAMP analog

Cell-permeable cAMP analog; activator of protein kinase A.

Chemische identificatiekenmerken

CAS 76939-46-3
Molecular Formula C10H11BrN5NaO6P
Molecular Weight (g/mol) 431.09
MDL Number MFCD00005844
InChI Key DMRMZQATXPQOTP-UHFFFAOYNA-N
Synonym 8-bromo-camp
PubChem CID 131954754
IUPAC Name sodium 6-(6-amino-8-bromo-9H-purin-9-yl)-2,7-dihydroxy-hexahydro-2λ⁵-furo[3,2-d][1,3,2]dioxaphosphinin-2-one
SMILES [Na+].NC1=C2N=C(Br)N(C3OC4COP(O)(=O)OC4C3O)C2=NC=N1

Specificatie

Quantity 50 mg
Formula Weight 430.09
Percent Purity >99%
Chemical Name or Material 8-Bromo-cAMP, sodium salt
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.