Learn More
Applied Biosystems™ 3500 Genetic Analyzer 8-Capillary Array, 50 cm
A radio-frequency identification tag is built into the frame for tracking critical run information, which is then reported on the software dashboard.
Brand: Applied Biosystems™ 4404685
1995.99 EUR valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Description
The internally uncoated capillaries are supplied in preassembled sets of 8 capillaries per array, with a built-in frame for easy installation.Order Info
Each
Specifications
8-Capillary Array | |
Store this array at ambient temperature. For details about storing the capillary array on and off the instrument, please refer to the Applied Biosystems 3500/3500xL Genetic Analyzer User Guide. | |
3500 Genetic Analyzer | |
125 min. | |
1 array |
50 cm | |
Protein Quality Analysis, Fragment Analysis, Sequencing | |
Room Temperature | |
3500 Genetic Analyzer 8-Capillary Array |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
L517F65 | Certificate of Analysis | 18/01/2018 | 4404685 |
L516H57 | Certificate of Analysis | 17/01/2018 | 4404685 |
L517I62 | Certificate of Analysis | 16/01/2018 | 4404685 |
L517H55 | Certificate of Analysis | 13/01/2018 | 4404685 |
L517H60 | Certificate of Analysis | 13/01/2018 | 4404685 |
Promotions
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24156Expires: 2025-06-30