Learn More
Mitutoyo™ 811 Series 10 to 90 Shore A Digital Display Durometers
Suitable digital Shore durometer for testing the hardness of a variety of rubber and natural elastomer materials. Mitutoyo™ 811 Series 10 to 90 Shore A Digital Display Durometers uses a Shore A scale for measurement.
Brand:  Mitutoyo™ 81133601
Encompass
Product Code. 11731604
  • 2355.00€ / Each
Only null left
Estimated Shipment: 24-06-2025
Add to basket
Includes: Data hold, zero-set, SPC data output
Description
Description
  • Suitable for testing the following materials:
    • natrual rubber, neoprene, polyesters, PVC, leather, Thiokol, nitrile rubber, wax, vinyl, cellulose acetates, glass polystyrene, etc.
  • Designed in accordance with ASTM D 2240, ISO 868, ISO 7619, DIN 53 505, JIS K 6253, and JIS K 7215 specifications.
  • Includes a data hold function for error-free reading from the LCD screen.
Specifications
Specifications
LCD
SR44
10 to 90 Shore A
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Mitutoyo™ 811 Series 10 to 90 Shore A Digital Display Durometers