Learn More
Uvex™ 8845 Hi-Vis Warning Protection Winter Parka
Shop All Uvex Products
Click to view available options
2X-Large
Description
- Functional, high-visibility parka with reflectors and thermal padding
- Certified according to: EN ISO 20471 X3, EN 343 (3/1)
- Reflective strips on the torso and sleeves, according to EN 471
- Concealed 2-way front zip with chin guard
- 2 side pockets, 2 breast pockets, cell phone pocket, pen pocket, and inside pocket
- Contrasting colored inserts on the shoulder and the front side
- Padded, firmly integrated lining for thermal insulation
- Adjustable hem and waist
Specifications
Specifications
Size | 2X-Large |
Gender | Unisex |
Material | Polyester |
Color | Hi-vis Yellow |
Closure Type | Zipper |
No. of Pockets | 7 |
Product Type | Hi-vis Jacket |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Special Offers
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Product Title
Uvex™ 8845 Hi-Vis Warning Protection Winter Parka >