Learn More
Eppendorf™ 96-channel Dispensing Heads
For use with epMotion™ 96 Flex in hard-shell case. Requires base device for operation.
Brand: Eppendorf™ 5069000535
6250.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- For use with epMotion™ 96 Flex (not included)
- Intuitive semi-automated handling of 96- and 384-well plates
- Calibrated according to ISO 23783-2 standards
- Includes hard shell case for transport and storage
Specifications
Dispensing Head | |
5 to 1000 ÎĽL | |
epMotion™ 96 Flex, and epT.I.P.S.™ Motion Reload System | |
1 Ea. | |
2.7 kg |
Not Autoclavable | |
Cell culture, Molecular biology | |
Air-cushion system | |
6.0 lb. | |
1 Ea. |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 7
Videos
-
epMotion 96 Flex tutorial Change of dispensing heads
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
2Hours
15Minutes
41Seconds
42
Product Content Correction
Product Title
Eppendorf™ 96-channel Dispensing Heads > 5 to 1000 μL