Learn More
Greiner Bio-One 96-well Chimney designed Round (U) Bottom Microplates
For high-throughput screening in cell based test systems.
Brand: Greiner Bio-One 650209
This item is not returnable.
View return policy
Description
- Non-pyrogenic
- Conical well design with flat bottom
- Ensures accurate pipetting
- Reduces dead volume of well
- Free of detectable DNase, RNase, human DNA
- Prevents wicking
Specifications
No | |
Without Lid | |
Not Autoclavable | |
Black | |
96 | |
Non-treated | |
Round Bottom (U-well) | |
100 | |
Yes | |
Chimney |
Standard | |
8 x 12 | |
Non Binding | |
Polypropylene | |
Non-sterile | |
355 μL | |
4800 x g (swinging bucket rotor) | |
100/Cs. | |
Black |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Greiner Bio-One 96-well Chimney designed Round (U) Bottom Microplates > Black