Learn More
Greiner Bio-One 96-well Chimney Style, Non-binding Microplates with μClear™ Film Bottom
To significantly improve data quality and sensitivity in biochemical assays. Greiner Bio-One 96-Well, Chimney Style Non-binding Microplates with Clear™ Film Bottom are free of detectable DNase, RNase, human DNA.
Brand: Greiner Bio-One 655906
Additional Details : Weight : 0.80000kg
Description
- Reduces non-specific biomolecule binding (proteins, DNA, RNA) for biochemical assays
- Stable under normal assay conditions without degradation or leaching
- Improves signal-to-noise-ratio
- Manufactured under DIN ISO 9001 guidelines
Service Plan
No
Services
No
Specifications
No | |
Without Lid | |
Non Binding | |
Polystyrene | |
Non-sterile | |
392 μL | |
40 | |
40/Cs. | |
127.8 x 85.5 x 14.4 mm |
Standard | |
8 x 12 | |
Black | |
96 | |
Non-treated | |
Flat Bottom (F-well) | |
10 | |
Clear |