missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One 96-well High Binding Standard ELISA Microplates
For diagnostics and immunological research applications. Greiner Bio-One 96-Well High Binding Standard ELISA Microplates is free of detectable DNase, RNase, human DNA.
Brand: Greiner Bio-One 650061
This item is not returnable.
View return policy
Description
- Non-Pyrogenic
- Free of detectable DNase, RNase, human DNA
- Material: Crystal clear polystyrene
- Alphanumeric well coding
- Suitable for transmission measurements
- Well-to-well spacing: 9mm
- Specifically tre
Service Plan
No
Certifications
DIN ISO 9001
Services
No
Specifications
Standard | |
High Binding | |
Polystyrene | |
Non-sterile | |
Square, Round Bottom (U-well) | |
Round |
8 x 12 | |
Clear | |
96 | |
Microlon 600 | |
40/Cs. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Greiner Bio-One 96-well High Binding Standard ELISA Microplates > Sterile; U-bottom (round)
Spot an opportunity for improvement?Share a Content Correction