Learn More
IKA™ A 11.6 Double Beater
Surface-hardened titanium beater with two blades for use in a grinding chamber.
Brand:  IKA™ 0003302900
Product Code. 10478511
  • 574.00€ / Each
Estimated Shipment: 24-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use with items up to Mohs hardness 3.
  • Only applicable with grinding chamber A 11.4.
  • Surface-hardened titanium construction.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Double Beaker
Materials with Mohs hardness up to 3
A 11.4 Grinding chamber
Surface-hardened Titanium
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
IKA™ A 11.6 Double Beater > For use with: A 11.4 Grinding chamber