Learn More
Tocris Bioscience™ A12B4C3
Potent and selective polynucleotide kinase/phosphatase (PNKP) inhibitor
Brand:  Tocris Bioscience™ 5097/10
277.31 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18710118
  • 351.00€ / 10mg
Estimated Shipment: 13-11-2024
Description
A12B4C3 is a potent and selective polynucleotide kinase/phosphatase (PNKP) inhibitor (IC50 = 60 nM). Displays no effect on calcineurin, protein PP-1 or APTX. Interrupts DNA repair and enhances radiosensitivity of human A549 lung carcinoma and MDA-MB-231 cells.
Specifications
Specifications
>98%
A12B4C3
A12B4C3
10 mg
534.65
Certificates
Ofertas especiales
Ofertas especiales
For Research Use Only