Learn More
Tocris Bioscience™ A12B4C3
Potent and selective polynucleotide kinase/phosphatase (PNKP) inhibitor
Brand:  Tocris Bioscience™ 5097/50
1090.26 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18728788
  • 1380.00€ / 50mg
Estimated Shipment: 13-11-2024
Description
A12B4C3 is a potent and selective polynucleotide kinase/phosphatase (PNKP) inhibitor (IC50 = 60 nM). Displays no effect on calcineurin, protein PP-1 or APTX. Interrupts DNA repair and enhances radiosensitivity of human A549 lung carcinoma and MDA-MB-231 cells.
Specifications
Specifications
>98%
A12B4C3
A12B4C3
50 mg
534.65