Learn More
A37, Tocris Bioscience™
Product Code. 30001024
Click to view available options
Quantity:
10 mg
50 mg
ALDH1A1 inhibitor
ALDH1A1 inhibitor (IC50 = 4.6 μM; Ki = 300 nM). Exhibits selectivity for ALDH1A1 over eight other ALDH isoforms. Disrupts spheroid formation and reduces cell viability of ovarian cancer cells in vitro.
Chemical Identifiers
CAS 896795-60-1
Molecular Formula C21H25N3O3S2
Molecular Weight (g/mol) 431.569
InChI Key SKDRHRAYBYQVNU-UHFFFAOYSA-N
Synonym CM 037
PubChem CID 5049161
IUPAC Name ethyl 2-[[4-oxo-3-(3-pyrrolidin-1-ylpropyl)-[1]benzothiolo[3,2-d]pyrimidin-2-yl]sulfanyl]acetate
SMILES CCOC(=O)CSC1=NC2=C(C(=O)N1CCCN3CCCC3)SC4=CC=CC=C42
Specifications
Linear Formula C21H25N3O3S2
Quantity 50 mg
Formula Weight 431.57
Physical Form Solid
Chemical Name or Material Ethyl 2-[[3,4-dihydro-4-oxo-3-[3-(1-pyrrolidinyl)propyl][1]benzothieno[3,2-d]pyrimidin-2-yl]thio]ace
Grade >97%