missing translation for 'onlineSavingsMsg'
Learn More

AAK1 293T Cell Overexpression Lysate (Denatured), Abnova

Brand:  Abnova H00022848-T01.100uL


Product Code. 16156652

  • 215.00€ / 100µL
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Western Blotting

Specifications

Specifications

293T
AAK1 293T Cell Transient Overexpression Lysate(Denatured)
52.25
pCMV-AAK1 full-length
100 μL
Human
Western Blot
BC002695.2
6.8
Transient overexpression cell Lysate was tested with Anti-AAK1 antibody (H00022848-B01) by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 100UL AAK1 293T Cell Transient Overexpression Lysa

Signal Word
  • Warning

Hazard Category
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

Supplemental information
  • MIXTURE LIST-Contains : Mercaptoethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.