missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Abiraterone acetate

Product Code. 17231272
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 154229-18-2
Molecular Formula C26H33NO2

Specifications

Quantity 250 mg
Melting Point 147.0°C
Formula Weight 391.55
InChI Key UVIQSJCZCSLXRZ-HMMZIKKISA-N
SMILES CC(=O)O[C@H]1CC[C@]2(C)C3CC[C@@]4(C)C(CC=C4C4=CC=CN=C4)C3CC=C2C1
IUPAC Name (7S,9aR,11aS)-9a,11a-dimethyl-1-(pyridin-3-yl)-3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-yl acetate
Molecular Weight (g/mol) 391.56
Chemical Name or Material Abiraterone acetate
compliance-icons compliance-icons
Product Identifier
  • Abiraterone acetate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Abiraterone acetate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'