Learn More
Abnova™ Fluorescent Dye 594-I Succinimidyl Ester
Water-soluble fluorescent dye forms stable peptide bond with protein amino groups
Brand: Abnova™ U0245.1mg
Description
Biomolecules conjugated to Fluorescent Dye 594-I Succinimidyl Ester exhibit little spectral overlap with green-fluorescent conjugates, and can be efficiently excited by 568nm line of Ar-Kr laser and by the 594nm line of orange He-Ne laser. Ideal for labeling of biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis
- Spectral Equivalent: Alexa Fluor 594, DyLight 594, Texas Red
- Purity: ≥90%
- Molar Extinction Co-efficient: >80,000 M-1cm-1
- Excitation (Max): 592nm
- Emission (Max): 614nm
- Solubility: DMSO, water
- Storage Buffer: DMSO
Application: Conjugation
Recommended Storage: Store in desiccated environment at -20°C and avoid from light.
Specifications
Fluorescent dye that is water-soluble and forms stable peptide bond with protein amino groups. Biomolecules conjugated to Fluorescent Dye 594-I Succinimidyl Ester exhibit little spectral overlap with green-fluorescent conjugates, and can be efficiently excited by 568nm line of Ar-Kr laser and by the 594nm line of orange He-Ne laser. | |
1 mg | |
≥90% | |
DMSO | |
Fluorescent Dye 594-I Succinimidyl Ester | |
592 |
Conjugation | |
Fluorescent Dye 594-I Succinimidyl Ester | |
Alexa Fluor 594, DyLight 594, Texas Red | |
Store the product in desiccated environment at -20°C and avoid from light. | |
614 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For research use only (RUO).