Learn More
Novus Biologicals™ AbSelect™ G Antibody Purification Kit
For purification of antibodies from crude samples such as ascites fluid or immune serum and removal of contaminants
Brand: Novus Biologicals™ 890-0010
This item is not returnable.
View return policy
Description
The method involves capturing the antibody on the AbSelect Protein G resin. Protein G has a high affinity for the Fc regions of IgG molecules from a variety of species and once the antibody has bound to the Protein G, unwanted substances can be removed by simply washing the resin. The purified product is then eluted and neutralized.
- Compatible with the lightning-link and lightning-link rapid conjugation systems
- Allows purified antibody to be immediately labeled with a hands-on time of under 30 seconds
Recommended Storage
Store at 4°C. Do not freeze.
Specifications
Antibody Purification Kit | |
3 Purifications | |
4°C. Do not freeze. | |
3 Tests |