Learn More
Fisherbrand™ Accessories for Fisherbrand™ Heating/Cooling Mini Shaker
Optional accessories to enhance the Fisher Scientific Heating/Cooling Mini Shaker
Brand:  Fisherbrand™ 88861104
82.56 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
 View more versions of this product
Product Code. 15584080
  • 99.70€ / Each
Estimated Shipment: 02-09-2025
Specifications
20 x 1.5 mL micro centrifuge tube block; Well: 11.1mm dia X 39.1mm depth
Special Offers
  • Fisherbrand
    Get up to 60% off select Fisherbrand products
    Promo Code: 24162
    Expires2025-03-31
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2025 and 31/03/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions Save Now
  • Multiple Manufacturers
    Browse exclusive prices for our academic customers
    Promo Code: 25081
    Expires2025-12-31
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
Product Content Correction
Product Title
Fisherbrand™ Accessories for Fisherbrand™ Heating/Cooling Mini Shaker > 20 x 1.5mL micro centrifuge tube block; Well: 11.1mm dia X 39.1mm depth