Learn More
Fisherbrand™ Accessories for Fisherbrand™ Multi-Purpose Tube Rotator
Optional accessories to enhance the Fisherbrand™Multi-Purpose Tube Rotator
Brand:  Fisherbrand™ 88861124
120.27 EUR valid until 2025-03-31
Use promo code "24162" to get your promotional price.
 View more versions of this product
Product Code. 15584090
  • 147.00€ / Each
In Stock
to see stock.
Edge
Description
Multi-purpose tube rotator offers flexibility with multiple carousel options
  • Tube rotator
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10 and 20 place tube carousel for 10-15mL and 5-7mL tubes
Special Offers
  • Multiple Manufacturers
    Browse exclusive prices for our academic customers
    Promo Code: 25081
    Expires2025-12-31
    New Offer
    Save Now
  • Multiple Manufacturers
    Explore exclusive monthly deals on essential consumables for unbeatable savings.
    Promo Code: 24222
    Expires2025-03-31
  • Fisherbrand
    Get up to 60% off select Fisherbrand products
    Promo Code: 24162
    Expires2025-03-31
    New Offer
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2025 and 31/03/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions Save Now
Product Content Correction
Product Title
Fisherbrand™ Accessories for Fisherbrand™ Multi-Purpose Tube Rotator > 10 and 20 place tube carousel for 10-15mL and 5-7mL tubes