accessibility menu, dialog, popup
Test
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
Learn More
Home
Products
pH and Electrochemistry
pH and Electrochemistry Meter Accessories
Benchtop Titrator Accessories
Mettler Toledo™ Accessories for G20 Titrator Models
Print
Mettler Toledo™ Accessories for G20 Titrator Models
For use with G20 Titrator models.
227.60€
Products
1
Documents
Product Certifications
Products
1
Product Code
Brand
Type
Price
Quantity & Availability
Product Code
Brand
Type
Price
Quantity & Availability
12705222
View Documents
missing translation for 'viewProductCertificates'
Mettler Toledo™
00089601
Cable
227.60€
Each
Estimated Shipment: 13-01-2025
Log in
Description
For use with G20 Titrator models.
Documents
Product Certifications
Back to Top