Documents
Scientific Resources
Application Notes
- 1
- 2
Obtain an excellent combination of speed and quality of separation using these solid core columns that are optimized for LC-MS and UHPLC-MS methods.
Brand: Thermo Scientific™ 17626-104630
459.16 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
757.42 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
Optimized for MS detection, Thermo Scientific™ Accucore™ RP-MS LC Columns are an excellent combination of speed and quality of separation.
Based on Core Enhanced Technology, Accucore columns provide fast, high-resolution separations—without the elevated backpressures required by sub-2μm particles.
Ordering Information:
Other custom column dimensions are available. Please call your local Customer Service for more information.
Analytical Column | |
HPLC | |
14500 psi (1000 bar) | |
2.6 ÎĽm | |
80 Ă… | |
60°C | |
Proprietary | |
Spherical, Solid Core, Ultrapure Silica | |
Reversed Phase | |
Reversed Phase |
4.6 mm | |
100 mm | |
Spherical Solid Core | |
2 to 9 | |
130 m²/g | |
N/A | |
Accucore | |
7% | |
Yes |
Expires: 2025-03-31