Learn More
Acetone oxime, 98%, Thermo Scientific Chemicals Product Code.: 11489722
Acetone oxime, 98%, Thermo Scientific Chemicals
Product Code. 11489722
1000 g
Click to view available options
Quantity:
50 g
250 g
1000 g
Product Code. 11489722
Brand: Thermo Scientific Alfa Aesar A10802.0B
Request Bulk or Custom Format
CAS: 127-06-0 | C3H7NO | 73.10 g/mol
It is an intermediate used in organic synthesis and in agriculture. It is also an important raw material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an intermediate used in organic synthesis and in agriculture. It is also an important raw material.
Solubility
Soluble in water (330 g/L at 20°C).
Notes
Store away from oxidizing agents. Protect against electrostatic charges.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 127-06-0
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00002118
InChI Key PXAJQJMDEXJWFB-UHFFFAOYSA-N
Synonym acetone oxime, acetoxime, propan-2-one oxime, 2-propanone, oxime, 2-propanone oxime, acetonoxime, acetone, oxime, beta-isonitrosopropane, acetoneoxime, ccris 5
PubChem CID 67180
ChEBI CHEBI:15349
IUPAC Name N-propan-2-ylidenehydroxylamine
SMILES CC(C)=NO
Specifications
Melting Point 60°C to 63°C
Density 0.901
Boiling Point 133°C to 135°C
Flash Point 60°C (140°F)
Quantity 1000 g
UN Number UN1325
Beilstein 1560146
Merck Index 14,75
Solubility Information Soluble in water (330g/L at 20°C).
Formula Weight 73.1
Percent Purity 98%
Chemical Name or Material Acetone oxime
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10254551 Certificate TypeCertificate of Analysis Date17/05/2025 Catalog NumberA10802.30,A10802.0B,A10802.18
Lot Number10242961 Certificate TypeCertificate of Analysis Date29/08/2022 Catalog NumberA10802.30,A10802.0B,A10802.18
Lot Number10186442 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10802.30,A10802.0B,A10802.18
Lot NumberD9945B Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10802.30,A10802.0B,A10802.18
Lot Number10155445 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10802.30,A10802.0B,A10802.18
Product Title
Acetone oxime, 98%, Thermo Scientific Chemicals >