Learn More
Olympus™ Achromatic Objective, SZX-ACH 1X
Replace old or worn your objectives with quality Olympus parts. Olympus™ Achromatic Objective, SZX-ACH 1X fits SZX-series microscopes.
Brand: Olympus™ K10017142
This item is not returnable.
View return policy
Description
- Achromatic
Specifications
Achromat Objective | |
SZX microscopes | |
0.1 |
WD 90mm | |
1X |