missing translation for 'onlineSavingsMsg'
Learn More

Acifran, Tocris Bioscience™

Product Code. 30001452
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Hypolipidemic agent; agonist for the GPR109A (HM74A) and GPR109B receptors

Hypolipidemic agent. Exhibits higher potency than nicotinic acid and clofibrate. Full and potent agonist at the human orphan GPCR HM74A/GPR109A and GPR109B (EC50 values are 1.3 and 4.2 μM respectively). Reduces serum triglycerides and circulating LDL-cholesterol in vivo, without affecting liver weight or liver enzymes.

Chemical Identifiers

CAS 72420-38-3
Molecular Formula C12H10O4
Molecular Weight (g/mol) 218.208
InChI Key DFDGRKNOFOJBAJ-UHFFFAOYSA-N
Synonym acifran, reductol, acifran usan:inn, acifranum inn-latin, unii-fp3llw01bl, unii-61iz92gn57, fp3llw01bl, 2-furancarboxylic acid, 4,5-dihydro-5-methyl-4-oxo-5-phenyl, 4,5-dihydro-5-methyl-4-oxo-5-phenyl-2-furancarboxylic acid, +--4,5-dihydro-5-methyl-4-oxo-5-phenyl-2-furoic acid
PubChem CID 51576
IUPAC Name 5-methyl-4-oxo-5-phenylfuran-2-carboxylic acid
SMILES CC1(C(=O)C=C(O1)C(=O)O)C2=CC=CC=C2

Specifications

Quantity 10 mg
Formula Weight 218.21
Percent Purity >99%
Chemical Name or Material Acifran
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.