missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acipimox, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Specifications
Quantity | 1 g |
Solubility | Soluble in methanol |
Merck Index | 13-113 |
Melting Point | 180°C to 190°C |
Formula Weight | 154.12 |
Physical Form | Powder |
Color | White to Almost-white |
Purity | ≥98.0% |
InChI Key | DNRXJHATQULEHC-UHFFFAOYSA-N |
SMILES | CC1=CN=CC(C(O)=O)=[N+]1[O-] |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction