Learn More
Cytiva AcroPak™ 200 Capsules with Supor™ Membrane
AcroPak capsules designed for research applications only.
Brand: Cytiva 12941
320.15 EUR valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Description
- Efficient processing of liquid volumes up to 20 L
- Built-in prefilter extends filter life when particulate-laden solutions, such as serum-containing media, are processed
- For buffers, tissue culture media, and other biological fluids
- Newer technology recommended for simplified scale-up
- High filtration area and compact size are ideal for upscale trials
Specifications
Hydrophilic PES | |
Sterile | |
Stepped Hose Barb |
0.8/0.2 ÎĽm | |
200 cm2 | |
3/Pk. |
Promotions
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24156Expires: 2025-06-30
Days
32Hours
19Minutes
53Seconds
15
Product Content Correction
Product Title
Cytiva AcroPak™ 200 Capsules with Supor™ Membrane > Stepped Hose Barb