missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Actinomycin D, 93%

Product Code. 10308880
25 mg, Glass Bottle
Click to view available options
Quantity:
1 mg
5 mg
25 mg
100 mg
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-76-0
Molecular Formula C62H86N12O16
Molecular Weight (g/mol) 1255.438
MDL Number MFCD00005033
InChI Key RJURFGZVJUQBHK-HUZQGMAJSA-N
Synonym Dactinomycin
PubChem CID 131954673
IUPAC Name 2-amino-4,6-dimethyl-3-oxo-1-N-[(3R,6S,7R,10S,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,11,14-tetrazabicyclo[14.3.0]nonadecan-6-yl]-9-N-[(3R,6S,7R,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,
SMILES CC1C(C(=O)NC(C(=O)N2CCCC2C(=O)N(CC(=O)N(C(C(=O)O1)C(C)C)C)C)C(C)C)NC(=O)C3=C4C(=C(C=C3)C)OC5=C(C(=O)C(=C(C5=N4)C(=O)NC6C(OC(=O)C(N(C(=O)CN(C(=O)C7CCCN7C(=O)C(NC6=O)C(C)C)C)C)C(C)C)C)N)C

Specifications

Type Actinomycin D
Physical Form Crystalline Powder
CAS Min % 91.0
CAS Max % 100.0
Melting Point 245.0°C to 252.0°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Color Red
Quantity 25 mg
Assay Percent Range 93%
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol, 1,2-propanediol, water + glyco, l mixture
Formula Weight 1255.44
Percent Purity 93%
Chemical Name or Material Actinomycin D, 93%
Show More Show Less
compliance-icons
Product Identifier
  • Actinomycin D
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
Hazard Statement
  • H300-Fatal if swallowed.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Actinomycin D, 93% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.