Learn More
Carl Zeiss™ Adapters
83.70€ - 1100.00€
Products 7
Product Code Brand Type Price Quantity & Availability  
Product Code Brand Type Price Quantity & Availability  
10738683
View Documents
Carl Zeiss™
456140-0000-000
Preparation Piece 60-44
144.00€
Each
Estimated Shipment: 11-06-2025
Add to basket
11891083
View Documents
Carl Zeiss™
452995-0000-000
Video Adapter 44 C 2/3, 1,0X This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
11876482
View Documents
Carl Zeiss™
452997-0000-000
Video Adapter 44 C, 0.66 in., 0.63X This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
11810903
View Documents
Carl Zeiss™
452998-0000-000
Video Adapter 44, 0.5 in., 0.5X This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
12574346
View Documents
Carl Zeiss™
426103-0000-000
Adapter 60N - T2, 1.0X
83.70€
Each
Estimated Shipment: 11-06-2025
Add to basket
11866472
View Documents
Carl Zeiss™
000000-1069-415
Video Adapter 60 C, 0.5 in., 0.5X
943.00€
Each
Estimated Shipment: 18-06-2025
Add to basket
11855143
View Documents
Carl Zeiss™
415500-1825-000
DSLR P95-T2 1.6x Adapter
1100.00€
Each
Estimated Shipment: 11-06-2025
Documents
Product Certifications
Product Content Correction
Product Title
Carl Zeiss™ Adapters