Learn More
Eppendorf™ Adaptor
Brand:  Eppendorf™ 4960800066
13.20 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10053231
Specifications
Adapter
Bottle thread
1 Ea.
38 mm
Documents
One moment while we fetch your results.
Product Title
Eppendorf™ Adaptor > Thread Size: 38mm