Learn More
Thermo Scientific Chemicals Adenosine-5'-diphosphate disodium salt, 96% (dry wt.), water 15% max. Product Code.: 11308905
Thermo Scientific Chemicals Adenosine-5'-diphosphate disodium salt, 96% (dry wt.), water 15% max.
Product Code. 11308905
1 g
Click to view available options
Quantity:
250 mg
1 g
Product Code. 11308905
Brand: Thermo Scientific Chemicals L14029.03
Request Bulk or Custom Format
Adenosine-5'-diphosphate disodium salt is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Adenosine-5′-diphosphate disodium salt is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is -20°C. Store under inert gas. Moisture sensitive. It is hygroscopic in nature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 16178-48-6
Molecular Formula C10H13N5Na2O10P2
Molecular Weight (g/mol) 471.17
MDL Number MFCD00150927
InChI Key ORKSTPSQHZNDSC-WCYUCLFNNA-L
Synonym adenosine 5'-diphosphate disodium salt, adenosine-5'-diphosphate disodium salt, sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl hydrogendiphosphate, adenosine 5'-diphosphate disodium salt hydrate, adenosine, 5'-diphosphoric acid, disodium salt, adenosine 5'-trihydrogen diphosphate , disodium salt, trinosin s hydrate, disodium hydrogen 2r,3s,4r,5r-5-6-aminopurin-9-yl-3,4-dihydroxyoxolan-2-yl methyl phosphonato oxyphosphonate, adenosine 5'-trihydrogen diphosphate , sodium salt 1:2, disodium adp hydrateShow MoreShow Less
PubChem CID 85315
IUPAC Name disodium;[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] hydrogen phosphate
SMILES [Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1
Specifications
Quantity 1 g
Beilstein 3645656
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Optical Rotation −30° (c=0.5 in 0.5M Na2HPO4)
Formula Weight 471.17
Percent Purity 97%
Assay (dry wt.), water ≤7%
Chemical Name or Material Adenosine-5'-diphosphate disodium salt
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252246 Certificate TypeCertificate of Analysis Date09/09/2024 Catalog NumberL14029.03,L14029.MD
Lot Number10250422 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberL14029.03,L14029.MD
Lot Number10237793 Certificate TypeCertificate of Analysis Date24/02/2023 Catalog NumberL14029.03,L14029.MD
Lot Number10242163 Certificate TypeCertificate of Analysis Date25/08/2022 Catalog NumberL14029.03,L14029.MD
Lot Number10215118 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL14029.03,L14029.MD
Product Title
Thermo Scientific Chemicals Adenosine-5'-diphosphate disodium salt, 96% (dry wt.), water 15% max. >