missing translation for 'onlineSavingsMsg'
Learn More

Adenosine A2aR Antibody (599717) [Janelia Fluor™ 549], Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals FAB9497I

Product Code. 18578467

  • 620.00€ / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Adenosine A2aR Monoclonal antibody specifically detects Adenosine A2aR in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Adenosine A2aR
Monoclonal
Janelia Fluor 549
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Flow Cytometry
599717
Flow Cytometry
adenosine A2 receptor, adenosine A2a receptor, adenosine receptor A2a, adenosine receptor subtype A2a, ADORA 2, ADORA2, hA2aR, RDC8
NS0 mouse myeloma cell line transfected with human Adenosine A2a R, Met1-Ser412, Accession # P29274
0.1 mL
Apoptosis, GPCR, Immunology, Innate Immunity
135
Store at 4°C in the dark.
IgG2a
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Adenosine A2aR Antibody (599717) [Janelia Fluor™ 549], Novus Biologicals™ > 0.1 mL; Janelia Fluor 549

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.