Learn More
Eppendorf™ Aerosol-Tight Cap, Universal Buckets, S-4xUniversal Rotor
Seals centrifuge rotor buckets for protection against hazardous materials.
Brand: Eppendorf™ 5910750005
227.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Suitable for S-4xUniversal Rotor buckets
- Creates aerosol-tight seal when working with hazardous substances
Specifications
S-4xUniversal rotor buckets |
Promotions
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
-
Multiple Manufacturers
Cancer Research Campaign
Save big on essential products for cancer researchPromo Code: 24147Expires: 2025-07-31
New Offer Save Now -
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
New Offer Save Up to 59%