Learn More
Invitrogen™ Shrimp Alkaline Phosphatase (SAP)
Treat unincorporated dNTPs in PCR reactions to prepare templates for DNA sequencing or SNP analysis. Shrimp Alkaline Phosphatase (SAP) is useful in molecular biology applications such as the dephosphorylation of phosphorylated ends of DNA or RNA for subsequent use in cloning or end-labeling of probes.
POA
Specifications
For Use With (Application) Dephosphorylation of DNA/RNA
View More Specs
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15533687
View Documents Promotion Details
Invitrogen™
78390500UN
500 U
POA
500 units
15523687
View Documents Promotion Details
Invitrogen™
783901000UN
1000 U
POA
1000 units
15543687
View Documents Promotion Details
Invitrogen™
783905000UN
5000 U
POA
5000 units
Description
Description
  • 100% heat-inactivated in 5 min at 65°C
  • Significantly improved storage stability at lower temperatures
  • Very high specific activity (see Fig. 3)
  • Removes 5’-phosphates from DNA, RNA, dNTPs, and proteins
  • Purified from a recombinant source
  • May be added directly to restriction enzyme digests
  • No vector purification necessary
  • Requires no supplemental zinc or other additives for activity
  • Works direct in many different buffers
  • Easy treatment of unincorporated dNTPs in PCR products prior to DNA sequencing or SNP analysis
Specifications
Specifications
Dephosphorylation of DNA/RNA
Documents
Product Certifications
Product Content Correction
Product Title
Invitrogen™ Shrimp Alkaline Phosphatase (SAP)