Learn More
Agarose SERVA™ Premium, Serva Electrophoresis™
Click to view available options
Quantity:
100 g
250 g
Specifications
Specifications
Physical Form | Powder |
Melting Point | 88.3°C |
Composition | Agarose SERVA Premium |
Density | 0.5 at 20°Cg/cm3 |
Color | White |
pH | 6 |
Quantity | 250 g |
Electroendosmosis (EEO) | 0.05 to 0.13 |
Gel Strength | >1100g/cm2 |
Solubility Information | 15 g/L |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 7
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Agarose SERVA™ Premium, Serva Electrophoresis™