Learn More
Giese™ Agate Pestle
Brand:  Giese™ 600-P-080
 View more versions of this product
Product Code. 11796794
  • 58.30€ / Each
Estimated Shipment: 09-05-2025
Specifications
80 mm
Product Title
Giese™ Agate Pestle > Length: 80mm