Learn More
Giese™ Agate Pestle
Brand:  Giese™ 600-P-130
 View more versions of this product
Product Code. 11833360
  • 383.00€ / Each
Estimated Shipment: 06-05-2025
Specifications
130 mm
Product Title
Giese™ Agate Pestle > Length: 130mm