Learn More
Brady™ Aggressive Adhesive Multipurpose Polyester Labels
Easily label components with the thermal transfer, glossy permanent polyester label material (B-422). Print almost anywhere with a durable Brady™ handheld label printer.
Brand: Brady™ 170739
Description
- Designed for a wide range of applications such as labeling equipment, circuit boards, panel components and barcodes
- Constructed to withstand abrasions, extreme temperatures and solvents such as alcohol, oil and water
- Made with a permanent acrylic adhesive that sticks to rough, smooth and powder-coated flat surfaces
Specifications
UL Recognised;CSA Accepted Compliant with Directive 2011/65/EU and 2015/863 | |
Permanent Acrylic | |
Gloss | |
Polyester | |
Non-sterile | |
BMP41;BMP51;BMP53;M511;M410;M510 | |
-40°F to 212°F | |
0.004 in | |
25.40 mm (W) x 25.40 mm (H) | |
1 in | |
1 in |
M4-49-422 | |
White | |
Die-cut | |
Labels | |
Asset tracking;Barcode labelling;Component and equipment labelling;Data and telecommunications labelling;General identification;Inventory and inspection labelling | |
260 | |
-40°C to 100°C | |
0.102 mm | |
1 in. | |
25.40 mm | |
25.40 mm |