Learn More
Gilac™ Airtight Box with White Lid
Airtight Box with White Lid is ideal as delivery box and click and collect service.
53.05€ - 88.55€
Specifications
Closure Type | Airtight Lid |
---|---|
Color | White |
Material | HDPE |
Shape | Rectangular |
Product Type | Airtight Box |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
17958901
|
Gilac™
G602012 |
0.6 L |
53.05€
Each |
Estimated Shipment: 27-03-2025
Log in to see stock. |
||||||
17918511
|
Gilac™
G602112 |
1.2 L |
88.55€
Each |
In Stock
Log in to see stock. ![]() |
||||||
17958831
|
Gilac™
G602212 |
2.5 L |
79.65€
Each |
In Stock
Log in to see stock. ![]() |
||||||
Description
- Airtight container, by pack of 12, perfect as delivery box and click and collect service.
- Keeps flavours in and odours out.
- Clear box: contents are visible and recognizable.
- Flexible, watertight lid.
- PP clear container. LDPE lid
- Available in 3 sizes, stackable.
- Customisation available on request for a minimum of 500 units.
Specifications
Airtight Lid | |
HDPE | |
Airtight Box | |
3 in. |
White | |
Rectangular | |
75 mm |
Product Title
Gilac™ Airtight Box with White Lid