missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Insulating Tube, Both Ends Open (Thin Wall);OD (mm), 1;ID (mm), 0.6, Thermo Scientific Chemicals

17.34€ - 49.85€

Specifications

Chemical Name or Material Al-23 Insulating Tube
Name Note Both ends open (Thin Wall)
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11347488
View Documents Promotion Details
Thermo Scientific Alfa Aesar
031479.DM
100 mm
17.34€
100mm
Estimated Shipment: 26-11-2024
to see stock.
Add to basket
 
11357488
View Documents Promotion Details
Thermo Scientific Alfa Aesar
031479.DR
200 mm
33.37€
200mm
In Stock
to see stock.
Add to basket
 
11337488
View Documents Promotion Details
Thermo Scientific Alfa Aesar
031479.DW
300 mm
49.85€
300mm
Estimated Shipment: 26-11-2024
to see stock.
Add to basket
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Al-23 Insulating Tube
Both ends open (Thin Wall)
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : U

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only