Learn More
Al-23 Tube, Both Ends Open, O.D.: 4mm, I.D.: 2mm, Thermo Scientific Chemicals
30.20€ - 141.00€
Specifications
Chemical Name or Material Al-23 Tube
Name Note Both ends open
View More Specs
Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11383917
View Documents
Thermo Scientific Alfa Aesar
031554.DM
100 mm This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
11343917
View Documents
Thermo Scientific Alfa Aesar
031554.DR
200 mm This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
11373917
View Documents
Thermo Scientific Alfa Aesar
031554.DW
300 mm
67.94€
300mm
In Stock
to see stock.
Add to basket
11353917
View Documents
Thermo Scientific Alfa Aesar
031554.EH
600 mm This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
11363917
View Documents
Thermo Scientific Alfa Aesar
031554.EM
800 mm
141.00€
800mm
Estimated Shipment: 22-01-2025
to see stock.
Add to basket
11333917
View Documents
Thermo Scientific Alfa Aesar
031554.EO
1000 mm This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Al-23 Tube
Both ends open
Videos
Safety and Handling
Safety and Handling
missing translation for 'tsca' : U
Recommended Storage : Ambient temperatures