Learn More
Al-23 Tube, Both ends open (Thin walls), O.D.: 6mm, I.D.: 5mm, Thermo Scientific Chemicals
POA - 62.89€
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11318247
|
Thermo Scientific Alfa Aesar
031706.DM |
100 mm |
35.10€
100mm |
Estimated Shipment: 28-10-2024
Log in to see stock. |
|||||
11308247
|
Thermo Scientific Alfa Aesar
031706.DR |
200 mm |
53.30€
200mm |
Estimated Shipment: 28-10-2024
Log in to see stock. |
|||||
11388237
|
Thermo Scientific Alfa Aesar
031706.DW |
300 mm | |||||||
15451237
|
Thermo Scientific Alfa Aesar
031706.Q@ |
400 mm | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Both ends open (Thin walls) |
Safety and Handling
missing translation for 'tsca' : U
Recommended Storage : Ambient temperatures
RUO – Research Use Only