Learn More
Al-24 Tube, Both ends open, O.D.: 30mm, I.D.: 24mm, Thermo Scientific Chemicals
42.49€ - 364.00€
Specifications
Chemical Name or Material Al-24 Tube
Name Note Both ends open
View More Specs
Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11394107
View Documents Promotion Details
Thermo Scientific Alfa Aesar
031606.DM
100 mm This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
Save Up to 
»
11344117
View Documents Promotion Details
Thermo Scientific Alfa Aesar
031606.DR
200 mm This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
Save Up to 
»
11324117
View Documents Promotion Details
Thermo Scientific Alfa Aesar
031606.DW
300 mm This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
Save Up to 
»
11334117
View Documents Promotion Details
Thermo Scientific Alfa Aesar
031606.EH
600 mm
251.00€
600mm
View alternative products
Save Up to 
»
Estimated Shipment: 28-10-2024
to see stock.
Add to basket
11304117
View Documents Promotion Details
Thermo Scientific Alfa Aesar
031606.EM
800 mm This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
Save Up to 
»
11314117
View Documents Promotion Details
Thermo Scientific Alfa Aesar
031606.EO
1000 mm
364.00€
1000mm
View alternative products
Save Up to 
»
In Stock
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Al-24 Tube
Both ends open
Videos
Safety and Handling
Safety and Handling
missing translation for 'tsca' : U
Recommended Storage : Ambient temperatures