missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Albumin bovine, for biochemistry, Fraction V, pH=5, 96.0-100.0% protein Product Code.: 10326640

Thermo Scientific Chemicals Albumin bovine, for biochemistry, Fraction V, pH=5, 96.0-100.0% protein

Product Code. 10326640
100 g
Click to view available options
Quantity:
10 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 10326640

Brand: Thermo Scientific Chemicals 134731000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Albumin, bovine serum (BSA), CAS # 9048-46-8, is a small, stable biomolecule derived from bovine blood with wide applications in biochemistry, immunochemistry, pharmacokinetics, and cell culture studies.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Albumin, bovine serum (BSA) is available as a protease-free, lyophilized powder that may be stably stored at 2-8°c
  • It is comprised of 583 amino acid residues, and has a molecular weight of 66.5 kDa

Applications

  • BSA is used as a protein standard in quantification assays, such as the Bradford protein assay
  • It is used to stabilize enzymes in restriction digestions in molecular biology experiments
  • It is a blocking agent in immunoblots thus increasing signal to noise ratios by quenching non-specific immunochemical reactions
  • BSA is a nutritive agent and used as a supplement in cell and microbial culture media
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9048-46-8

Specifications

Packaging Glass bottle
Quantity 100 g
Solubility Information Solubility aq. soln.: soluble.
Grade Biochemistry
Chemical Name or Material Albumin bovine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466176 Certificate TypeCertificate of Analysis Date25/09/2024 Catalog Number134735000,134731000,134730100
Lot NumberA0460855 Certificate TypeCertificate of Analysis Date18/03/2024 Catalog Number134735000,134731000,134730100
Lot NumberA0455085 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number134735000,134731000,134730100
Lot NumberA0449737 Certificate TypeCertificate of Analysis Date30/01/2023 Catalog Number134735000,134731000,134730100
Lot NumberA0441020 Certificate TypeCertificate of Analysis Date11/04/2022 Catalog Number134735000,134731000,134730100
Product Identifier
  • Albumin bovine / fraction V, pH=5, 96.0-100.0% protein

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Albumin bovine, for biochemistry, Fraction V, pH=5, 96.0-100.0% protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.