Learn More
Albumin, bovine, Thermo Scientific Chemicals
Brand: Thermo Scientific J30867.A1
Description
Specifications
1 kg | |
Plastic bottle | |
Lyophilized |
Keep cold | |
Albumin |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
182420 | Certificate of Analysis | 27/11/2022 | J30867.A1 |
185238 | Certificate of Analysis | 27/11/2022 | J30867.A1 |
185061 | Certificate of Analysis | 27/11/2022 | J30867.A1 |
221472 | Certificate of Analysis | 23/06/2022 | J30867.A1 |
221908 | Certificate of Analysis | 23/06/2022 | J30867.A1 |