Learn More
Novus Biologicals™ Albumin Native Protein
Native Protein
Brand:  Novus Biologicals™ NBP1-97049-5mg
Product Code. 18462841
  • 534.00€ / 5mg
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Add to basket
Specifications
P02768
Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
5 mg
albumin, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin
Reconstitute with 500 μL of deionzied water.
Apoptosis, Cancer, Cellular Markers, Golgi Apparatus Markers, Stem Cell Markers
Albumin
Dot Blot, PAGE
213
Store at 4°C.
ALB
This product has been tested in SDS-Page and can be used in SDS, Western Blotting, ELISA experiments and other immunological assays.
Multi-step